missing translation for 'onlineSavingsMsg'
Learn More

N-(n-Butyl)thiourea, 98%

Catalog No. p-7045162
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0965404 2 g
AAL0965409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0965404 Supplier Thermo Scientific Chemicals Supplier No. L0965404

CAS: 1516-32-1 | C5H12N2S | 132.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1516-32-1
Molecular Formula C5H12N2S
Molecular Weight (g/mol) 132.23
MDL Number MFCD00022173
InChI Key GMEGXJPUFRVCPX-UHFFFAOYSA-N
Synonym 1-butyl-2-thiourea, n-butylthiourea, thiourea, butyl, n-butyl thiourea, butyl-2-thiourea, thiourea, n-butyl, usaf d-5, 1-butylthiourea, urea, 1-butyl-2-thio, n-n-butyl thiourea
PubChem CID 1551919
IUPAC Name butylthiourea
SMILES CCCCNC(N)=S

Specifications

Melting Point 76°C to 79°C
Odor Odorless
Quantity 2 g
Beilstein 1744779
Formula Weight 132.23
Percent Purity 98%
Chemical Name or Material N-(n-Butyl)thiourea
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 216-165-9
RTECSNumber YS4375000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only