accessibility menu, dialog, popup

UserName

CAS RN 1516-32-1

IUPAC Name: butylthiourea
Synonyms: thiourea, butyl-
Molecular Weight (g/mol): 132.23
Molecular Formula: C5H12N2S
InChi Key: GMEGXJPUFRVCPX-UHFFFAOYSA-N
SMILES: CCCCNC(N)=S
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)