missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(trimethylsilyl)urea 98.0+%, TCI America™

Catalog No. B110325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B110325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B110325G Supplier TCI America Supplier No. B110325G

Chemical Identifiers

CAS 18297-63-7
Molecular Formula C7H20N2OSi2
Molecular Weight (g/mol) 204.42
MDL Number MFCD00008260
InChI Key MASDFXZJIDNRTR-UHFFFAOYSA-N
Synonym 1,3-bis trimethylsilyl urea, n,n'-bis trimethylsilyl urea, bis trimethylsilyl urea, urea, n,n'-bis trimethylsilyl, n,n'-bis-trimethylsilyl-urea, urea, 1,3-bis trimethylsilyl, bsu, urea, bis-tms, hexamethyl disilaurea
PubChem CID 87562
IUPAC Name 1,3-bis(trimethylsilyl)urea
SMILES C[Si](C)(C)NC(=O)N[Si](C)(C)C

Specifications

Color White
Quantity 25 g
Formula Weight 204.42
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(trimethylsilyl)urea
EINECSNumber (2)-3250
TSCA Yes