missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Bis[4-(diphenylamino)phenyl]-N,N'-di(1-naphthyl)benzidine 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 5 g |
| Formula Weight | 923.18 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N,N′-Bis[4-(diphenylamino)phenyl]-N,N′-di(1-naphthyl)benzidine |
Safety and Handling
| TSCA | No |