missing translation for 'onlineSavingsMsg'
Learn More

N,N,3,5-Tetramethylaniline, 98%

Catalog No. AAL0926606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0926606 5 g
AAL0926614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0926606 Supplier Thermo Scientific Chemicals Supplier No. L0926606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4913-13-7 | C10H15N | 149.237 g/mol

N,N,3,5-Tetramethylaniline was used in rapid quenching of 1,8-dihydroxyanthraquinone (DHAQ). It was also used as a catalyst in the polymeric synthesis of glycol methacrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N,3,5-Tetramethylaniline was used in rapid quenching of 1,8-dihydroxyanthraquinone (DHAQ). It was also used as a catalyst in the polymeric synthesis of glycol methacrylate.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 4913-13-7
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00008307
InChI Key NBFRQCOZERNGEX-UHFFFAOYSA-N
Synonym benzenamine, n,n,3,5-tetramethyl, n,n-dimethyl-3,5-xylidine, 3,5-xylidine, n,n-dimethyl, 3,5-dimethylphenyl dimethylamine, acmc-209kew, 5-dimethylamino-m-xylene, 3,5,n,n-tetramethylaniline, n,n-dimethyl-sym-m-xylidine, n,n,3,5-tetramethyl aniline, n,n-3,5-tetramethylbenzenamine
PubChem CID 78625
IUPAC Name N,N,3,5-tetramethylaniline
SMILES CC1=CC(=CC(=C1)N(C)C)C

Specifications

Density 0.913
Quantity 5 g
Odor Recognizable
Refractive Index 1.544
UN Number UN2810
Beilstein 1933736
Boiling Point 226°C to 228°C
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 149.24
Percent Purity 98%
Chemical Name or Material N,N,3,5-Tetramethylaniline
Show More Show Less
Hazard Category H227-H315-H319-H335-H373
Hazard Statement GHS H Statement
H311-H373-H227-H302-H332
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 225-539-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only