missing translation for 'onlineSavingsMsg'
Learn More

N,N,2,4,6-Pentamethylaniline 97.0+%, TCI America™

Catalog No. P17675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17675G 5 g
P176725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17675G Supplier TCI America Supplier No. P17675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13021-15-3
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00010596
InChI Key JZBZLRKFJWQZHU-UHFFFAOYSA-N
Synonym n,n,2,4,6-pentamethylbenzenamine, 2-dimethylamino mesitylene, n,n-dimethylmesidine, benzenamine,n,n,2,4,6-pentamethyl, 2-dimethylaminomesitylene, dimethylaminomesitylen, pubchem3219, acmc-209bhq, n,n-2,4,6-pentamethylaniline, 2,4,6,n,n-pentamethylbenzenamine
PubChem CID 139365
IUPAC Name N,N,2,4,6-pentamethylaniline
SMILES CC1=CC(=C(C(=C1)C)N(C)C)C

Specifications

Color Yellow
Boiling Point 215°C
Quantity 5 g
Formula Weight 163.26
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N,2,4,6-Pentamethylaniline