missing translation for 'onlineSavingsMsg'
Learn More

N,N,2,4,6-Pentamethylaniline, 98%, Thermo Scientific Chemicals

Catalog No. AAA1056114
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1056114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1056114 Supplier Thermo Scientific Chemicals Supplier No. A1056114

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13021-15-3 | Molecular Formula: C11H17N | Molecular Weight: 163.264 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13021-15-3
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00010596
InChI Key JZBZLRKFJWQZHU-UHFFFAOYSA-N
Synonym n,n,2,4,6-pentamethylbenzenamine, 2-dimethylamino mesitylene, n,n-dimethylmesidine, benzenamine,n,n,2,4,6-pentamethyl, 2-dimethylaminomesitylene, dimethylaminomesitylen, pubchem3219, acmc-209bhq, n,n-2,4,6-pentamethylaniline, 2,4,6,n,n-pentamethylbenzenamine
PubChem CID 139365
IUPAC Name N,N,2,4,6-pentamethylaniline
SMILES CC1=CC(=C(C(=C1)C)N(C)C)C

Specifications

Density 0.907
Boiling Point 213°C to 215°C
Flash Point 78°C (174°F)
Refractive Index 1.521
Quantity 25 g
Formula Weight 163.27
Percent Purity 98%
Chemical Name or Material N,N,2,4,6-Pentamethylaniline
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only