missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpyrrolidone, B&J Brand™, for HPLC, GC and spectrophotometry, >99.5%, Honeywell Burdick & Jackson

Catalog No. 6003104
Change view
Click to view available options
Quantity:
20 L
Packaging:
Hdpe Nowpak™
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6003104 20 L Hdpe Nowpak™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6003104 Supplier Honeywell Chemicals Supplier No. NP30420
Only null left

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
Synonym 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
PubChem CID 13387
ChEBI CHEBI:7307
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

Absorbance 0.5 AU max. at 300 nm, 0.1 AU max. at 325 nm, 0.03 AU max. at 350 nm, 0.01 AU max. at 400 nm
CAS Max % 100.0000%
Density 1.0259 g/cm3
Boiling Point 202°C
Flash Point 88°C
Packaging Hdpe Nowpak™
Quantity 20 L
Beilstein 106420
Solubility Information Miscible in Water at 20°C
Formula Weight 99.15
Percent Purity >99.5%
Grade HPLC
Chemical Name or Material N-Methylpyrrolidone, B&J Brand™, for HPLC, GC and spectrophotometry, >99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Packing Group: 3