missing translation for 'onlineSavingsMsg'
Learn More

N-Methylisobutylamine 98.0+%, TCI America™

Catalog No. M069125ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M069125ML 25 mL
M0691100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M069125ML Supplier TCI America Supplier No. M069125ML

Chemical Identifiers

CAS 625-43-4
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00015043
InChI Key QKYWADPCTHTJHQ-UHFFFAOYSA-N
Synonym n-methylisobutylamine, 1-propanamine, n,2-dimethyl, n,2-dimethylpropylamine, n-isobutylmethylamine, methyl 2-methylpropyl amine, isobutylmethylamine, methylisobutylamine, propylamine, n,2-dimethyl, n-isobutyl-n-methylamine, methyl-isobutyl-amin
PubChem CID 12249
IUPAC Name N,2-dimethylpropan-1-amine
SMILES CC(C)CNC

Specifications

Color Colorless
Boiling Point 78°C
Quantity 25 mL
UN Number 2733
Formula Weight 87.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methylisobutylamine