missing translation for 'onlineSavingsMsg'
Learn More

N-Methyldiphenylamine 98.0+%, TCI America™

Catalog No. M02115ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M02115ML 5 mL
M021125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M02115ML Supplier TCI America Supplier No. M02115ML

Chemical Identifiers

CAS 552-82-9
Molecular Formula C13H13N
Molecular Weight (g/mol) 183.25
MDL Number MFCD00041900
InChI Key DYFFAVRFJWYYQO-UHFFFAOYSA-N
Synonym n-methyldiphenylamine, benzenamine, n-methyl-n-phenyl, methyldiphenylamine, n,n-diphenylmethylamine, diphenylamine, n-methyl, unii-b28zgh99ih, diphenyl methylamine, n-methyl-n-phenyl-aniline, b28zgh99ih, methyldiphenylamin
PubChem CID 11098
IUPAC Name N-methyl-N-phenylaniline
SMILES CN(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 295°C
Quantity 5 mL
Formula Weight 183.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methyldiphenylamine
EINECSNumber (3)-0364
TSCA Yes