missing translation for 'onlineSavingsMsg'
Learn More

N-Methylcyclohexylamine 99.0+%, TCI America™

Catalog No. M0202500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0202500ML 500 mL
M020225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0202500ML Supplier TCI America Supplier No. M0202500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 100-60-7
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.20
MDL Number MFCD00003832
InChI Key XTUVJUMINZSXGF-UHFFFAOYSA-N
Synonym n-methylcyclohexylamine, methylcyclohexylamine, n-cyclohexylmethylamine, cyclohexanamine, n-methyl, n-methyl-n-cyclohexylamine, methylamino cyclohexane, cyclohexylamine, n-methyl, cyclohexyl methyl amine, n-cyclohexyl-n-methylamine, unii-yh8pf4c1ow
PubChem CID 7514
ChEBI CHEBI:59018
IUPAC Name N-methylcyclohexanamine
SMILES CNC1CCCCC1

Specifications

Color Colorless
Boiling Point 148°C
Quantity 500 mL
UN Number 2734
Formula Weight 113.20
Percent Purity ≥99.0% (T)
Physical Form Liquid
Chemical Name or Material N-Methylcyclohexylamine
EINECSNumber (3)-4003
RTECSNumber GX1529000
TSCA Yes