missing translation for 'onlineSavingsMsg'
Learn More

N-Méthyl-N-triméthylsilylacétamide 90,0+%, TCI America™

Catalog No. M053625G
Change view
Click to view available options
Quantité:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
M053625G 25 g
M053610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M053625G Supplier TCI America Supplier No. M053625G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7449-74-3
Formule moléculaire C6H15NOSi
Poids moléculaire (g/mol) 145.277
Numéro MDL MFCD00040758
Clé InChI QHUOBLDKFGCVCG-UHFFFAOYSA-N
Synonyme n-methyl-n-trimethylsilyl acetamide, acetamide, n-methyl-n-trimethylsilyl, n-trimethylsilyl-n-methylacetamide, n-methyl-n-tms-acetamide, acmc-1bei6, qhuobldkfgcvcg-uhfffaoysa, n-methylacetamide, tms derivative, n-methyl-n-trimethylsilylacetamide, for gc derivatization gc
PubChem CID 81953
Nom de l’IUPAC N-méthyl-N-triméthylsilylacétamide
SOURIRES CC(=O)N(C)[Si](C)(C)C

Specifications

Nom ou substance chimique N-Methyl-N-trimethylsilylacetamide [Trimethylsilylating Agent]
Couleur Jaune
Point d’ébullition 154 °C
Quantité 25 g
Numéro UN 2924
Poids de formule 145.28
Pourcentage de pureté ≥95,0% (GC)
Forme physique Liquide
TSCA Oui
Recommended Storage Réfrigérateur