missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-(trifluoromethylthio)aniline 98.0+%, TCI America™

Catalog No. M2595200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2595200MG 200 mg
M25951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2595200MG Supplier TCI America Supplier No. M2595200MG
Only null left

Chemical Identifiers

CAS 66476-44-6
Molecular Formula C8H8F3NS
Molecular Weight (g/mol) 207.214
InChI Key TYFGNILTFVMYGX-UHFFFAOYSA-N
PubChem CID 25013449
IUPAC Name N-methyl-N-(trifluoromethylsulfanyl)aniline
SMILES CN(C1=CC=CC=C1)SC(F)(F)F

Specifications

Color Yellow
Quantity 200 mg
UN Number 1993
Formula Weight 207.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methyl-N-(trifluoromethylthio)aniline
TSCA No
Recommended Storage Refrigerator