missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-propargylbenzylamine 98.0+%, TCI America™

Catalog No. M26181G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M26181G Supplier TCI America Supplier No. M26181G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 555-57-7
Color Yellow
Boiling Point 90°C
Molecular Formula C11H13N
Quantity 1 g
MDL Number MFCD00008576
UN Number 2810
Synonym pargyline, pargylamine, n-methyl-n-propargylbenzylamine, paragyline, eudatin, supirdyl, eutonyl, pargylin, n-methyl-n-2-propynylbenzylamine, benzyl-methyl-2-propinylamin
InChI Key DPWPWRLQFGFJFI-UHFFFAOYSA-N
SMILES CN(CC#C)CC1=CC=CC=C1
IUPAC Name N-benzyl-N-methylprop-2-yn-1-amine
Molecular Weight (g/mol) 159.232
PubChem CID 4688
ChEBI CHEBI:7930
Formula Weight 159.23
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Methyl-N-propargylbenzylamine
Show More Show Less
RTECSNumber DP6475000
TSCA No