missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-((4-methyl-2-phenyl-1,3-thiazol-5-yl)methyl)amine, 97%, Thermo Scientific™

Numéro de catalogue. cc29246cb
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC29246CB 250mg
CC29246DA 1g
CC29246DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. CC29246CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC29246CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 850375-02-9
Formule moléculaire C12H14N2S
Poids moléculaire (g/mol) 218.318
Numéro MDL MFCD07368511
Clé InChI GFAGRBRYZWAUSV-UHFFFAOYSA-N
Synonyme n-methyl-n-4-methyl-2-phenyl-1,3-thiazol-5-yl methyl amine, n-methyl-1-4-methyl-2-phenyl-1,3-thiazol-5-yl methanamine, methyl 4-methyl-2-phenyl-1,3-thiazol-5-yl methyl amine, 5-thiazolemethanamine,n,4-dimethyl-2-phenyl, n-methyl-1-4-methyl-2-phenylthiazol-5-yl methanamine, methyl 4-methyl-2-phenyl 1,3-thiazol-5-yl methyl amine, mmj
PubChem CID 4961253
Nom de l’IUPAC N-methyl-1-(4-methyl-2-phenyl-1,3-thiazol-5-yl)methanamine
SOURIRES CC1=C(SC(=N1)C2=CC=CC=C2)CNC

Spécifications

Nom ou substance chimique N-methyl-N-[(4-methyl-2-phenyl-1, 3-thiazol-5-yl)methyl]amine
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber glass bottle
Quantité 250mg
Poids de formule 218.32
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.