missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[4-(2-morpholin-4-ylethoxy)benzyl]amine, 95%, Thermo Scientific™

Chemical Identifiers

CAS 852180-77-9
Molecular Formula C14H22N2O2
Molecular Weight (g/mol) 250.342
MDL Number MFCD07368554
InChI Key CFDWJPMGCABITD-UHFFFAOYSA-N
Synonym n-methyl-n-4-2-morpholin-4-ylethoxy benzyl amine, methyl 4-2-morpholin-4-yl ethoxy phenyl methyl amine, benzenemethanamine,n-methyl-4-2-4-morpholinyl ethoxy, n-methyl-1-4-2-morpholin-4-ylethoxy phenyl methanamine, n-methyl-n-4-2-morpholin-4-ylethoxy benzylamine, n-methyl-1-4-2-morpholinoethoxy phenyl methanamine, methyl 4-2-morpholin-4-ylethoxy phenyl methyl amine
PubChem CID 7060587
IUPAC Name N-methyl-1-[4-(2-morpholin-4-ylethoxy)phenyl]methanamine
SMILES CNCC1=CC=C(C=C1)OCCN2CCOCC2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC42146CB
SDS View Documents
Thermo Scientific Chemicals
CC42146CB
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC42146DA
SDS View Documents
Thermo Scientific Chemicals
CC42146DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC42146DE
SDS View Documents
Thermo Scientific Chemicals
CC42146DE
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

852180-77-9
250.342
CFDWJPMGCABITD-UHFFFAOYSA-N
7060587
CNCC1=CC=C(C=C1)OCCN2CCOCC2
C14H22N2O2
MFCD07368554
n-methyl-n-4-2-morpholin-4-ylethoxy benzyl amine, methyl 4-2-morpholin-4-yl ethoxy phenyl methyl amine, benzenemethanamine,n-methyl-4-2-4-morpholinyl ethoxy, n-methyl-1-4-2-morpholin-4-ylethoxy phenyl methanamine, n-methyl-n-4-2-morpholin-4-ylethoxy benzylamine, n-methyl-1-4-2-morpholinoethoxy phenyl methanamine, methyl 4-2-morpholin-4-ylethoxy phenyl methyl amine
N-methyl-1-[4-(2-morpholin-4-ylethoxy)phenyl]methanamine

Specifications

852180-77-9
100.0
Amber glass bottle
MFCD07368554
n-methyl-n-4-2-morpholin-4-ylethoxy benzyl amine, methyl 4-2-morpholin-4-yl ethoxy phenyl methyl amine, benzenemethanamine,n-methyl-4-2-4-morpholinyl ethoxy, n-methyl-1-4-2-morpholin-4-ylethoxy phenyl methanamine, n-methyl-n-4-2-morpholin-4-ylethoxy benzylamine, n-methyl-1-4-2-morpholinoethoxy phenyl methanamine, methyl 4-2-morpholin-4-ylethoxy phenyl methyl amine
CNCC1=CC=C(C=C1)OCCN2CCOCC2
250.342
250.34
N-Methyl-N-[4-(2-morpholin-4-ylethoxy)benzyl]amine
94.5
95%
C14H22N2O2
250mg
CFDWJPMGCABITD-UHFFFAOYSA-N
N-methyl-1-[4-(2-morpholin-4-ylethoxy)phenyl]methanamine
7060587
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'