missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-N-(3-pyridin-4-ylbenzyl)amine, 97%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 95% |
| Packaging | Amber Glass Bottle |
| Quantity | 250 mg |
| Formula Weight | 198.27 |
| Percent Purity | 95% |
| Chemical Name or Material | N-Methyl-N-(3-pyridin-4-ylbenzyl)amine |