missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[3-(1H-1,2,4-triazol-1-yl)benzyl]amine 97+%, Thermo Scientific™

Catalog No. CC38446CB
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC38446CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC38446CB Supplier Thermo Scientific Chemicals Supplier No. CC38446CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 879896-43-2
Molecular Formula C10H12N4
Molecular Weight (g/mol) 188.234
MDL Number MFCD08435864
InChI Key LLGQTEDUTKNYIP-UHFFFAOYSA-N
Synonym n-methyl-n-3-1h-1,2,4-triazol-1-yl benzyl amine, methyl 3-1,2,4-triazol-1-yl phenyl methyl amine, benzenemethanamine,n-methyl-3-1h-1,2,4-triazol-1-yl, methyl 3-1,2,4-triazolyl phenyl methyl amine, n-methyl-n-3-1h-1,2,4-trazol-1-yl benzyl amne, methyl 3-1h-1,2,4-triazol-1-yl phenyl methyl amine, 1-3-1h-1,2,4-triazol-1-yl phenyl-n-methylmethanamine, n-methyl-1-3-1h-1,2,4-triazol-1-yl phenyl methanamine
PubChem CID 18525742
IUPAC Name N-methyl-1-[3-(1,2,4-triazol-1-yl)phenyl]methanamine
SMILES CNCC1=CC(=CC=C1)N2C=NC=N2

Specifications

Assay Percent Range 97+%
Quantity 250 mg
Formula Weight 188.23
Chemical Name or Material n-methyl-n-[3-(1h-1, 2, 4-triazol-1-yl)benzyl]amine