missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-D-aspartic Acid 98.0+%, TCI America™

Catalog No. M1360100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1360100MG 100 mg
M13601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1360100MG Supplier TCI America Supplier No. M1360100MG

Chemical Identifiers

CAS 6384-92-5
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00004226
InChI Key HOKKHZGPKSLGJE-GSVOUGTGSA-N
Synonym n-methyl-d-aspartic acid, nmda, n-methyl-d-aspartate, n-methylaspartate, d-aspartic acid, n-methyl, methyl aspartic acid, n-me-d-asp-oh, r-2-methylamino succinic acid, unii-1903b9q6pi, n methyl d aspartate
PubChem CID 22880
ChEBI CHEBI:31882
IUPAC Name (2R)-2-(methylamino)butanedioic acid
SMILES CNC(CC(=O)O)C(=O)O

Specifications

Melting Point 178°C
Color White
Quantity 100 mg
Formula Weight 147.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl-D-aspartic Acid
RTECSNumber CI9457000
TSCA No