missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-D-aspartic acid, 98%

Catalog No. AC329192500
Change view
Click to view available options
Quantity:
50 mg
250 mg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC329192500 250 mg Glass bottle
AC329190500 50 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC329192500 Supplier Thermo Scientific Chemicals Supplier No. 329192500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6384-92-5 | C5H9NO4 | 147.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6384-92-5
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.1
MDL Number MFCD00004226
InChI Key HOKKHZGPKSLGJE-GSVOUGTGSA-N
Synonym n-methyl-d-aspartic acid, nmda, n-methyl-d-aspartate, n-methylaspartate, d-aspartic acid, n-methyl, methyl aspartic acid, n-me-d-asp-oh, r-2-methylamino succinic acid, unii-1903b9q6pi, n methyl d aspartate
PubChem CID 22880
ChEBI CHEBI:31882
IUPAC Name (2R)-2-(methylamino)butanedioic acid
SMILES CNC(CC(=O)O)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 187°C to 192°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 250 mg
Merck Index 15, 6749
Specific Rotation Condition − 28.00 (20.00°C c=1,HCl 6N)
Specific Rotation -28
Solubility Information Solubility in water: slightly soluble in water. Other solubilities: soluble in dilute aqueous base,slightly soluble i
Formula Weight 147.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Methyl-D-aspartic acid, 98%
Show More Show Less
Hazard Category LIGHT SENSITIVE
RTECSNumber CI9457000

RUO – Research Use Only