missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-D-aspartic acid, 98+%

Catalog No. AAJ61361MF
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61361MF 50 mg
AAJ61361MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61361MF Supplier Thermo Scientific Chemicals Supplier No. J61361MF

CAS: 6384-92-5 | C5H9NO4 | 147.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6384-92-5
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00004226
InChI Key HOKKHZGPKSLGJE-GSVOUGTGSA-N
Synonym n-methyl-d-aspartic acid, nmda, n-methyl-d-aspartate, n-methylaspartate, d-aspartic acid, n-methyl, methyl aspartic acid, n-me-d-asp-oh, r-2-methylamino succinic acid, unii-1903b9q6pi, n methyl d aspartate
PubChem CID 22880
ChEBI CHEBI:31882
IUPAC Name (2R)-2-(methylamino)butanedioic acid
SMILES CNC(CC(=O)O)C(=O)O

Specifications

Melting Point 187°C to 192°C
Color White
Beilstein 1724431
Merck Index 14,6662
Quantity 50 mg
Formula Weight 147.13
Optical Rotation −15° (c=2 in Water)
Percent Purity ≥98%
Sensitivity Hygroscopic
Physical Form Powder
Chemical Name or Material N-Methyl-D-aspartic acid
Show More Show Less
RTECSNumber CI9457000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only