missing translation for 'onlineSavingsMsg'
Learn More

n-methyl-[5-(morpholinomethyl)thien-2-ylmethyl]amine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC73446DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC73446DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

893742-71-7
226.34
LJPRPFHZPNCEIQ-UHFFFAOYSA-N
20098901
CNCC1=CC=C(CN2CCOCC2)S1
C11H18N2OS
MFCD06803277
n-methyl-5-morpholinomethyl thien-2-ylmethyl amine, methyl 5-morpholin-4-ylmethyl thiophen-2-yl methyl amine, n-methyl-1-5-morpholin-4-ylmethyl thiophen-2-yl methylamine, 2-methylamino methyl-5-morpholin-4-ylmethyl thiophene, 2-thiophenemethanamine,n-methyl-5-4-morpholinylmethyl, methyl 5-morpholin-4-ylmethyl 2-thienyl methyl amine, n-methyl-1-5-morpholinomethyl thiophen-2-yl methanamine, n-methyl-1-5-morpholin-4-yl methyl thiophen-2-yl methanamine
N-methyl-1-[5-(morpholin-4-ylmethyl)thiophen-2-yl]methanamine

Specifications

N-methyl-[5-(morpholinomethyl)thien-2-ylmethyl]amine
96.5
97%
C11H18N2OS
MFCD06803277
LJPRPFHZPNCEIQ-UHFFFAOYSA-N
N-methyl-1-[5-(morpholin-4-ylmethyl)thiophen-2-yl]methanamine
20098901
97%
893742-71-7
100.0
Amber Glass Bottle
1g
n-methyl-5-morpholinomethyl thien-2-ylmethyl amine, methyl 5-morpholin-4-ylmethyl thiophen-2-yl methyl amine, n-methyl-1-5-morpholin-4-ylmethyl thiophen-2-yl methylamine, 2-methylamino methyl-5-morpholin-4-ylmethyl thiophene, 2-thiophenemethanamine,n-methyl-5-4-morpholinylmethyl, methyl 5-morpholin-4-ylmethyl 2-thienyl methyl amine, n-methyl-1-5-morpholinomethyl thiophen-2-yl methanamine, n-methyl-1-5-morpholin-4-yl methyl thiophen-2-yl methanamine
CNCC1=CC=C(CN2CCOCC2)S1
226.34
226.34
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents