missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-3-phenoxybenzylamine, 97%, Thermo Scientific™

Numéro de catalogue. CC57146CB
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC57146CB 250mg
CC57146DA 1g
CC57146DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. CC57146CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC57146CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 129535-78-0
Formule moléculaire C14H15NO
Poids moléculaire (g/mol) 213.28
Numéro MDL MFCD02089413
Clé InChI GDVLOOQWWBPGSV-UHFFFAOYSA-N
Synonyme n-methyl-3-phenoxybenzylamine, n-methyl-1-3-phenoxyphenyl methanamine, methyl 3-phenoxyphenyl methyl amine, n-methyl-3-phenoxybenzenemethanamine, benzenemethanamine, n-methyl-3-phenoxy, acmc-209bgx, benzenemethanamine,n-methyl-3-phenoxy
PubChem CID 10512764
Nom de l’IUPAC N-methyl-1-(3-phenoxyphenyl)methanamine
SOURIRES CNCC1=CC(=CC=C1)OC2=CC=CC=C2

Spécifications

Nom ou substance chimique N-Methyl-3-phenoxybenzylamine
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber glass bottle
Quantité 250mg
Poids de formule 213.28
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.