missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-2-(tetrahydropyran-4-yloxy)benzylamine, 95%, Thermo Scientific™

Identifiants chimiques

CAS 906352-71-4
Molecular Formula C13H19NO2
Molecular Weight (g/mol) 221.3
MDL Number MFCD09064962
InChI Key ZTMRIJRRCHAUNA-UHFFFAOYSA-N
Synonym n-methyl-2-tetrahydropyran-4-yloxy benzylamine, n-methyl-1-2-tetrahydro-2h-pyran-4-yl oxy phenyl methanamine, methyl 2-oxan-4-yloxy phenyl methyl amine, n-methyl-n-2-tetrahydro-2h-pyran-4-yloxy benzyl amine, n-methyl-1-2-oxan-4-yl oxy phenyl methanamine, methyl 2-2h-3,4,5,6-tetrahydropyran-4-yloxy phenyl methyl amine, n-methyl-1-2-tetrahydro-2h-pyran-4-yl ;oxy ;phenyl ;methanamine
PubChem CID 24229518
IUPAC Name N-methyl-1-[2-(oxan-4-yloxy)phenyl]methanamine
SMILES CNCC1=CC=CC=C1OC2CCOCC2
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
CC52246CB
FDS Afficher les documents
Thermo Scientific Chemicals
CC52246CB
250mg Amber Glass Bottle N/A
N/A
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
CC52246DA
FDS Afficher les documents
Thermo Scientific Chemicals
CC52246DA
1g Amber Glass Bottle N/A
N/A
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Description

Description

Spécifications

Identifiants chimiques

906352-71-4
221.3
ZTMRIJRRCHAUNA-UHFFFAOYSA-N
24229518
CNCC1=CC=CC=C1OC2CCOCC2
C13H19NO2
MFCD09064962
n-methyl-2-tetrahydropyran-4-yloxy benzylamine, n-methyl-1-2-tetrahydro-2h-pyran-4-yl oxy phenyl methanamine, methyl 2-oxan-4-yloxy phenyl methyl amine, n-methyl-n-2-tetrahydro-2h-pyran-4-yloxy benzyl amine, n-methyl-1-2-oxan-4-yl oxy phenyl methanamine, methyl 2-2h-3,4,5,6-tetrahydropyran-4-yloxy phenyl methyl amine, n-methyl-1-2-tetrahydro-2h-pyran-4-yl ;oxy ;phenyl ;methanamine
N-methyl-1-[2-(oxan-4-yloxy)phenyl]methanamine

Spécifications

906352-71-4
100.0
Amber Glass Bottle
MFCD09064962
n-methyl-2-tetrahydropyran-4-yloxy benzylamine, n-methyl-1-2-tetrahydro-2h-pyran-4-yl oxy phenyl methanamine, methyl 2-oxan-4-yloxy phenyl methyl amine, n-methyl-n-2-tetrahydro-2h-pyran-4-yloxy benzyl amine, n-methyl-1-2-oxan-4-yl oxy phenyl methanamine, methyl 2-2h-3,4,5,6-tetrahydropyran-4-yloxy phenyl methyl amine, n-methyl-1-2-tetrahydro-2h-pyran-4-yl ;oxy ;phenyl ;methanamine
CNCC1=CC=CC=C1OC2CCOCC2
221.3
221.3
N-Methyl-2-(tetrahydropyran-4-yloxy)benzylamine
94.5
95%
C13H19NO2
250mg
ZTMRIJRRCHAUNA-UHFFFAOYSA-N
N-methyl-1-[2-(oxan-4-yloxy)phenyl]methanamine
24229518
95%
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

FDS
missing translation for 'documents'

missing translation for 'documents'