missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Méthyl-2-phényléthhylamine 97,0+%, TCI America™
Spécifications
Spécifications
| Color | Jaune |
| Boiling Point | 205°C |
| Quantity | 25 g |
| Formula Weight | 135.21 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Liquide |
| Chemical Name or Material | N-Methyl-2-phenylethylamine |
Sécurité et manipulation
| RTECSNumber | SH9625000 |
| TSCA | No |