missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-2-nitroaniline, 98%

Catalog No. AAA1953822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1953822 100 g
AAA1953806 5 g
AAA1953814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1953822 Supplier Thermo Scientific Chemicals Supplier No. A1953822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 612-28-2 | C7H8N2O2 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 612-28-2
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007090
InChI Key KFBOUJZFFJDYTA-UHFFFAOYSA-N
Synonym n-methyl-o-nitroaniline, benzenamine, n-methyl-2-nitro, n-methyl 2-nitroaniline, o-methylamino nitrobenzene, aniline, n-methyl-o-nitro, 2-nitro-n-methylaniline, o-nitro-n-methylaniline, n-methyl-2-nitrobenzenamine, o-nitro-n-methyl aniline, methyl 2-nitrophenyl amine
PubChem CID 69157
IUPAC Name N-methyl-2-nitroaniline
SMILES CNC1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 33°C to 36°C
Flash Point >110°C (230°F)
Odor Characteristic
Quantity 100 g
UN Number UN2811
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material N-Methyl-2-nitroaniline
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 210-303-1
RTECSNumber BY5710000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only