missing translation for 'onlineSavingsMsg'
Learn More

N-methyl-(1H-benzimidazol-5-ylmethyl)amine dioxalate, 90%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC45996CB

Quantity 250mg
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC45996CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

910036-85-0
341.276
HDQRBNFLWRHBAA-UHFFFAOYSA-N
18525786
CNCC1=CC2=C(C=C1)N=CN2.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O
C13H15N3O8
MFCD09025842
n-methyl-1h-benzimidazol-5-ylmethyl amine dioxalate, 1h-1,3-benzodiazol-5-ylmethyl methyl amine; bis oxalic acid, n-methyl-1h-benzo d imidazol-5-ylmethyl amine dioxalate, oxalic acid-1-1h-benzimidazol-6-yl-n-methylmethanamine 2/1, 3h-1,3-benzodiazol-5-ylmethyl methyl amine; bis oxalic acid
1-(3H-benzimidazol-5-yl)-N-methylmethanamine;oxalic acid

Specifications

N-methyl-(1H-benzimidazol-5-ylmethyl)amine dioxalate
89.5
90%
C13H15N3O8
MFCD09025842
HDQRBNFLWRHBAA-UHFFFAOYSA-N
1-(3H-benzimidazol-5-yl)-N-methylmethanamine;oxalic acid
18525786
90%
910036-85-0
100.0
Amber Glass Bottle
250mg
n-methyl-1h-benzimidazol-5-ylmethyl amine dioxalate, 1h-1,3-benzodiazol-5-ylmethyl methyl amine; bis oxalic acid, n-methyl-1h-benzo d imidazol-5-ylmethyl amine dioxalate, oxalic acid-1-1h-benzimidazol-6-yl-n-methylmethanamine 2/1, 3h-1,3-benzodiazol-5-ylmethyl methyl amine; bis oxalic acid
CNCC1=CC2=C(C=C1)N=CN2.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O
341.276
341.27
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents