missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-1-naphthylmethylamine Hydrochloride 98.0+%, TCI America™

Catalog No. M09845G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09845G 5 g
M098425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M09845G Supplier TCI America Supplier No. M09845G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 65473-13-4
Molecular Formula C12H14ClN
Molecular Weight (g/mol) 207.701
MDL Number MFCD00012555
InChI Key BVJVHPKFDIYQOU-UHFFFAOYSA-N
Synonym n-methyl-1-naphthalenemethylamine hydrochloride, n-methyl-1-naphthalen-1-yl methanamine hydrochloride, n-methyl-1-naphthylmethylamine hydrochloride, n-methyl-n-naphthylmethylamine hydrochloride, unii-z8vrt9b309, methyl naphthalen-1-ylmethyl amine hydrochloride, n-methyl-1-naphthalenemethanamine hydrochloride, n-methyl-c-naphthalen-1-yl methanamine hydrochloride, 1-naphthalenemethanamine, n-methyl-, hydrochloride 1:1, 1-methylaminomethyl naphthalene hydrochloride
PubChem CID 16211748
IUPAC Name N-methyl-1-naphthalen-1-ylmethanamine;hydrochloride
SMILES CNCC1=CC=CC2=CC=CC=C21.Cl

Specifications

Melting Point 193°C
Color White-Yellow
Quantity 5 g
Formula Weight 207.70
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl-1-naphthylmethylamine Hydrochloride