missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-1,3-propanediamine, 99%, Thermo Scientific™

Catalog No. AC155570010
Change view
Click to view available options
1L; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC155570010 1L; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC155570010 Supplier Thermo Scientific Chemicals Supplier No. 155570010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 6291-84-5
CAS Min % 98.5
CAS Max % 100.0
Density 0.8400g/mL
Boiling Point 139°C to 141°C
Flash Point 35°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C4H12N2
Refractive Index 1.4468
Linear Formula CH3NH(CH2)3NH2
Quantity 1L
MDL Number MFCD00008209
UN Number 2922
Beilstein 04,I,419
Specific Gravity 0.84
Synonym n-methyl-1,3-propanediamine, n-methyl-1,3-diaminopropane, 3-methylamino propylamine, 1,3-propanediamine, n-methyl, n-methylpropane-1,3-diamine, 3-aminopropyl methylamine, n-methyltrimethylenediamine, 3-aminopropylmethylamine, n-methyl-1,3-propylenediamine, 1-amino-3-methylamino propane
Solubility Information Solubility in water: soluble
InChI Key QHJABUZHRJTCAR-UHFFFAOYSA-N
SMILES CNCCCN
IUPAC Name N'-methylpropane-1,3-diamine
Molecular Weight (g/mol) 88.15
PubChem CID 80511
Viscosity 1.6 mPa.s (20°C)
Formula Weight 88.15
Percent Purity 99%
Chemical Name or Material N-Methyl-1, 3-propanediamine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Fatal if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 228-544-6
RTECSNumber TX8242500
TSCA TSCA

RUO – Research Use Only