missing translation for 'onlineSavingsMsg'
Learn More

N-(Methoxymethyl)methacrylamide (stabilized with MEHQ) 85.0+%, TCI America™

Catalog No. M247825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M247825G 25 g
M2478500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M247825G Supplier TCI America Supplier No. M247825G

Chemical Identifiers

CAS 3644-12-0
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00035808
InChI Key YOZHLACIXDCHPV-UHFFFAOYSA-N
PubChem CID 77204
IUPAC Name N-(methoxymethyl)-2-methylprop-2-enamide
SMILES COCNC(=O)C(C)=C

Specifications

Color Yellow
Boiling Point 85°C
Quantity 25 g
Formula Weight 129.16
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(Methoxymethyl)methacrylamide (stabilized with MEHQ)
EINECSNumber (2)-1067
TSCA Yes