missing translation for 'onlineSavingsMsg'
Learn More

N-Methoxy-N-methylbenzamide 98.0+%, TCI America™

Catalog No. M26725G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26725G 5 g
M267225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26725G Supplier TCI America Supplier No. M26725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6919-61-5
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00075320
InChI Key UKERDACREYXSIV-UHFFFAOYSA-N
Synonym n-methyl-n-methoxybenzamide, benzamide, n-methoxy-n-methyl, n-methoxy-n-methyl-benzamide, methyl n-methylbenzohydroxamate, pubchem17496, acmc-1b5bp, n-methoxy-n-methylbenzamid, n-methoxy-n-methyl benzamide, ksc445c3n, n-methoxy-n-methylbenzamide
PubChem CID 569575
ChEBI CHEBI:59742
IUPAC Name N-methoxy-N-methylbenzamide
SMILES CN(C(=O)C1=CC=CC=C1)OC

Specifications

Color Colorless
Boiling Point 112°C
Quantity 5 g
Formula Weight 165.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methoxy-N-methylbenzamide