missing translation for 'onlineSavingsMsg'
Learn More

N-Methoxy-N-methylbenzamide, 98%, Thermo Scientific™

Catalog No. AC436560050
Change view
Click to view available options
Quantity:
5g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC436560050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC436560050 Supplier Thermo Scientific Chemicals Supplier No. 436560050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6919-61-5
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
InChI Key UKERDACREYXSIV-UHFFFAOYSA-N
Synonym n-methyl-n-methoxybenzamide, benzamide, n-methoxy-n-methyl, n-methoxy-n-methyl-benzamide, methyl n-methylbenzohydroxamate, pubchem17496, acmc-1b5bp, n-methoxy-n-methylbenzamid, n-methoxy-n-methyl benzamide, ksc445c3n, n-methoxy-n-methylbenzamide
PubChem CID 569575
ChEBI CHEBI:59742
IUPAC Name N-methoxy-N-methylbenzamide
SMILES CN(C(=O)C1=CC=CC=C1)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Boiling Point 85°C (1.0mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5g
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material N-Methoxy-N-methylbenzamide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only