missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropylphthalimide 98.0+%, TCI America™

Catalog No. I073825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I073825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I073825G Supplier TCI America Supplier No. I073825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 304-17-6
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
MDL Number MFCD00014582
InChI Key VPLDXHDOGVIETL-UHFFFAOYSA-N
PubChem CID 67535
IUPAC Name 2-(propan-2-yl)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES CC(C)N1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 84°C
Color White-Yellow
Boiling Point 273°C
Quantity 25 g
Formula Weight 189.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Isopropylphthalimide
EINECSNumber (5)-3353
RTECSNumber TI5425000
TSCA Yes