missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropylcyclohexylamine 98.0+%, TCI America™

Catalog No. I0373500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0373500ML 500 mL
I037325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0373500ML Supplier TCI America Supplier No. I0373500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1195-42-2
Molecular Formula C9H19N
Molecular Weight (g/mol) 141.258
MDL Number MFCD00003831
InChI Key UYYCVBASZNFFRX-UHFFFAOYSA-N
Synonym n-isopropylcyclohexylamine, cyclohexylisopropylamine, n-isopropylcyclohexanamine, cyclohexylamine, n-isopropyl, isopropylcyclohexylamine, cyclohexanamine, n-1-methylethyl, n-cyclohexylisopropylamine, n-cyclohexyl-n-isopropylamine, n-propan-2-yl cyclohexanamine, n-isopropyl cyclohexylamine
PubChem CID 62386
IUPAC Name N-propan-2-ylcyclohexanamine
SMILES CC(C)NC1CCCCC1

Specifications

Color Yellow
Boiling Point 174°C
Quantity 500 mL
UN Number 2734
Formula Weight 141.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Isopropylcyclohexylamine
RTECSNumber GX1450000
TSCA No