missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxysulfosuccinimide sodium salt, 95%

Catalog No. AC438650050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC438650050 5 g Glass bottle
AC438650010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC438650050 Supplier Thermo Scientific Chemicals Supplier No. 438650050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Coupling reagent | CAS: 106627-54-7 | C4H4NNaO6S | 217.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Chemical Identifiers

CAS 106627-54-7
Molecular Formula C4H4NNaO6S
Molecular Weight (g/mol) 217.13
InChI Key RPENMORRBUTCPR-UHFFFAOYSA-M
Synonym n-hydroxysulfosuccinimide sodium salt, sulfo-nhs, sodium 1-hydroxy-2,5-dioxopyrrolidine-3-sulfonate, n-hydroxysulfosuccinimidesodiumsalt, 3-pyrrolidinesulfonic acid, 1-hydroxy-2,5-dioxo-, monosodium salt, sodium 1-hydroxy-2,5-dioxo-pyrrolidine-3-sulfonate, n-hydroxusulfosuccinimide sodium, n-hydroxy-2-sodiosulfo succinimide, n-hydroxysulfosuccinimide,sodium salt, n-hydroxusulfosuccinimide sodium salt
PubChem CID 3520574
IUPAC Name sodium;1-hydroxy-2,5-dioxopyrrolidine-3-sulfonate
SMILES C1C(C(=O)N(C1=O)O)S(=O)(=O)[O-].[Na+]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 250.0°C
Color White
Elemental Analysis 21.46 to 23.02% C, 6.19 to 6.71% N
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 5 g
Formula Weight 217.13
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Hydroxysulfosuccinimide sodium salt
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
If skin ir

RUO – Research Use Only