missing translation for 'onlineSavingsMsg'
Learn More

n-Hexyltrimethoxysilane, 97%

Catalog No. AAL0357614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0357614 25 g
AAL0357606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0357614 Supplier Thermo Scientific Chemicals Supplier No. L0357614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3069-19-0 | C9H22O3Si | 206.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3069-19-0
Molecular Formula C9H22O3Si
Molecular Weight (g/mol) 206.36
MDL Number MFCD00039801
InChI Key CZWLNMOIEMTDJY-UHFFFAOYSA-N
Synonym n-hexyltrimethoxysilane, silane, hexyltrimethoxy, unii-9aui936hic, hexyl trimethoxy silane, 9aui936hic, hexyl-trimethoxysilane, acmc-209hhw, czwlnmoiemtdjy-uhfffaoysa
PubChem CID 76478
IUPAC Name hexyl(trimethoxy)silane
SMILES CCCCCC[Si](OC)(OC)OC

Specifications

Boiling Point 202°C to 203°C
Quantity 25 g
Flash Point 62°C (143°F)
Refractive Index 1.407
Sensitivity Moisture sensitive
Density 0.92
Beilstein 2347551
Formula Weight 206.36
Percent Purity 97%
Chemical Name or Material n-Hexyltrimethoxysilane
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
EINECSNumber 221-331-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only