missing translation for 'onlineSavingsMsg'
Learn More

n-Hexyl benzoate, 98+%

CAS: 6789-88-4 | C13H18O2 | 206.285 g/mol

Supplier:  Thermo Scientific Chemicals L0629322

 View more versions of this product

Catalog No. AAL0629322


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

n-Hexyl benzoate
0.98
148°C (298°F)
1.493
100 g
benzoic acid, hexyl ester, benzoic acid n-hexyl ester, n-hexylbenzoate, n-hexyl benzoate, 1-hexyl benzoate, hexylbenzoate, hexylester kyseliny benzoove, benzoic acid hexyl ester, unii-vr3c36ekjr, fema no. 3691
CCCCCCOC(=O)C1=CC=CC=C1
206.285
206.29
6789-88-4
272°C
C13H18O2
MFCD00051714
2048117
UUGLJVMIFJNVFH-UHFFFAOYSA-N
hexyl benzoate
23235
≥98%

Chemical Identifiers

6789-88-4
206.285
UUGLJVMIFJNVFH-UHFFFAOYSA-N
23235
CCCCCCOC(=O)C1=CC=CC=C1
C13H18O2
MFCD00051714
benzoic acid, hexyl ester, benzoic acid n-hexyl ester, n-hexylbenzoate, n-hexyl benzoate, 1-hexyl benzoate, hexylbenzoate, hexylester kyseliny benzoove, benzoic acid hexyl ester, unii-vr3c36ekjr, fema no. 3691
hexyl benzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 229-856-5

RTECSNumber : DH1490000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only