accessibility menu, dialog, popup

UserName

H3COO

CAS RN 6789-88-4

IUPAC Name: hexyl benzoate
Synonyms: hexyl benzoate
Molecular Weight (g/mol): 206.29
Molecular Formula: C13H18O2
InChi Key: UUGLJVMIFJNVFH-UHFFFAOYSA-N
SMILES: CCCCCCOC(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)