missing translation for 'onlineSavingsMsg'
Learn More

n-Hexyl benzoate, 98+%

Catalog No. p-7044413
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0629314 25 g
AAL0629322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0629314 Supplier Thermo Scientific Chemicals Supplier No. L0629314
Only null left

CAS: 6789-88-4 | C13H18O2 | 206.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6789-88-4
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.285
MDL Number MFCD00051714
InChI Key UUGLJVMIFJNVFH-UHFFFAOYSA-N
Synonym benzoic acid, hexyl ester, benzoic acid n-hexyl ester, n-hexylbenzoate, n-hexyl benzoate, 1-hexyl benzoate, hexylbenzoate, hexylester kyseliny benzoove, benzoic acid hexyl ester, unii-vr3c36ekjr, fema no. 3691
PubChem CID 23235
IUPAC Name hexyl benzoate
SMILES CCCCCCOC(=O)C1=CC=CC=C1

Specifications

Density 0.98
Boiling Point 272°C
Flash Point 148°C (298°F)
Refractive Index 1.493
Quantity 25 g
Beilstein 2048117
Formula Weight 206.29
Percent Purity ≥98%
Chemical Name or Material n-Hexyl benzoate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 229-856-5
RTECSNumber DH1490000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only