missing translation for 'onlineSavingsMsg'
Learn More

n-Heptyl 4-hydroxybenzoate, 98%

Catalog No. AAA1355514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1355514 25 g
AAA1355522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1355514 Supplier Thermo Scientific Chemicals Supplier No. A1355514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1085-12-7 | C14H20O3 | 236.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
n-Heptyl 4-hydroxybenzoate is used for inhibiting microbiological spoilage, used as preservatives, skin care, hair care.

Solubility
Soluble in alcohol, phenoxyethanol, propylene glycol. Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 1085-12-7
Molecular Formula C14H20O3
Molecular Weight (g/mol) 236.31
MDL Number MFCD00016481
InChI Key ZTJORNVITHUQJA-UHFFFAOYSA-N
Synonym heptylparaben, heptyl paraben, heptyl p-hydroxybenzoate, n-heptyl 4-hydroxybenzoate, nipaheptyl, staypro ws 7, n-heptyl p-hydroxybenzoate, benzoic acid, 4-hydroxy-, heptyl ester, p-hydroxybenzoic acid heptyl ester, p-oxybenzoesaureheptylester
PubChem CID 14138
ChEBI CHEBI:34788
IUPAC Name heptyl 4-hydroxybenzoate
SMILES CCCCCCCOC(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 46°C to 49°C
Quantity 25 g
Solubility Information Soluble in alcohol,phenoxyethanol,propylene glycol. Insoluble in water.
Formula Weight 236.31
Percent Purity 98%
Chemical Name or Material n-Heptyl 4-hydroxybenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-115-0
RTECSNumber DH2202000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only