missing translation for 'onlineSavingsMsg'
Learn More

N-Formylglycine 98.0+%, TCI America™

Catalog No. F01215G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F01215G 5 g
F012125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F01215G Supplier TCI America Supplier No. F01215G

Chemical Identifiers

CAS 2491-15-8
Molecular Formula C3H5NO3
Molecular Weight (g/mol) 103.08
MDL Number MFCD00037360
InChI Key UGJBHEZMOKVTIM-UHFFFAOYSA-N
Synonym n-formylglycine, formylglycine, glycine, n-formyl, fgly, for-gly-oh, formylglycin, n-hydroxymethyleneglycine, formylamino acetic acid, n-formylglycin, n-formyl-glycine
PubChem CID 75606
IUPAC Name 2-formamidoacetic acid
SMILES OC(=O)CNC=O

Specifications

Melting Point 147°C
Color White
Quantity 5 g
Formula Weight 103.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Formylglycine
RTECSNumber MC0547000
TSCA No