missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl-L-proline, 97%

Numéro de catalogue. p-7077617
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AC456420010 1 g
AC456420050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC456420010 Fournisseur Thermo Scientific Chemicals Code fournisseur 456420010
Il en reste null

CAS: 13200-83-4 | C6H9NO3 | 143.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 13200-83-4
Molecular Formula C6H9NO3
Molecular Weight (g/mol) 143.14
InChI Key DHDRGOURKDLAOT-YFKPBYRVSA-N
Synonym 1-formyl-l-proline, l-proline, 1-formyl, n-formylproline, n-formyl-l-proline, 2s-1-formylpyrrolidine-2-carboxylic acid, s---n-formylproline, n-formyl-l-proline tlc, s-1-formyl-2-pyrrolidinecarboxylic acid, s-1-formylpyrrolidine-2-carboxylic acid
PubChem CID 12461843
IUPAC Name (2S)-1-formylpyrrolidine-2-carboxylic acid
SMILES C1CC(N(C1)C=O)C(=O)O

Spécifications

Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Specific Rotation -125
Formula Weight 143.14
Percent Purity 97%
Chemical Name or Material N-Formyl-L-proline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 236-165-2

RUO – Research Use Only