missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-N-methyl-L-valine, 95%
Description
N-Fmoc-N-methyl-L-valine is used as enzyme substrates and reagents, culture media additives, dyes, stains and indicators. It is also involved in the preparation of N-methylated peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Fmoc-N-methyl-L-valine is used as enzyme substrates and reagents, culture media additives, dyes, stains and indicators. It is also involved in the preparation of N-methylated peptides.
Solubility
Slightly soluble in water and dimethyl formamide.
Notes
Incompatible with strong oxidizing agents, reducing agents, bases and alkali metals. Store in a cool place.
N-Fmoc-N-methyl-L-valine is used as enzyme substrates and reagents, culture media additives, dyes, stains and indicators. It is also involved in the preparation of N-methylated peptides.
Solubility
Slightly soluble in water and dimethyl formamide.
Notes
Incompatible with strong oxidizing agents, reducing agents, bases and alkali metals. Store in a cool place.
Specifications
Specifications
| Melting Point | 187°C to 190°C |
| Color | White |
| Solubility Information | Slightly soluble in water and dimethyl formamide. |
| Quantity | 5 g |
| Formula Weight | 353.41 |
| Percent Purity | 95% |
| Physical Form | Powder |
| Chemical Name or Material | N-Fmoc-N-methyl-L-valine |
Safety and Handling
| TSCA | No |
| storageNote1 | 2° to 8°C |
| Recommended Storage | Keep cold |
RUO – Research Use Only