missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-pentafluorophenylalanine, 95%

Catalog No. AAH52059MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52059MD 250 mg
AAH5205903 1 g
AAH5205906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52059MD Supplier Thermo Scientific Chemicals Supplier No. H52059MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 205526-32-5 | C24H16F5NO4 | 477.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 205526-32-5
Molecular Formula C24H16F5NO4
Molecular Weight (g/mol) 477.39
MDL Number MFCD00270612
InChI Key DLOGILOIJKBYKA-KRWDZBQOSA-N
Synonym fmoc-pentafluoro-l-phenylalanine, fmoc-phe f5-oh, fmoc-l-pentafluorophenylalanine, fmoc-l-pentafluorophe, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-perfluorophenyl propanoic acid, fmoc-3-pentafluorophenyl-l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-pentafluoro-l-phenylalanine, n-alpha-9-fluorenylmethyloxycarbonyl-l-pentafluoro-phenylalanine, s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-pentafluorophenyl-propionic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-pentafluorophenyl propanoic acid
PubChem CID 7020337
SMILES OC(=O)[C@H](CC1=C(F)C(F)=C(F)C(F)=C1F)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Quantity 250 mg
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(2,3,4,5,6-pentafluorophenyl)propanoic acid
Formula Weight 477.39
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-pentafluorophenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only