missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-beta-homovaline, 95%

Catalog No. AAH52182MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52182MD 250 mg
AAH5218203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52182MD Supplier Thermo Scientific Chemicals Supplier No. H52182MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 172695-33-9 | C21H23NO4 | 353.42 g/mol

N-Fmoc-L-beta-homovaline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-L-beta-homovaline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 172695-33-9
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.42
MDL Number MFCD01862853
InChI Key KFDMTAMWOIQTOB-LJQANCHMSA-N
Synonym fmoc-l-beta-homovaline, r-3-9h-fluoren-9-yl methoxy carbonyl amino-4-methylpentanoic acid, fmoc-beta-hoval-oh, r-n-fmoc-3-amino-4-methylpentanoic acid, r-3-fmoc-amino-4-methylpentanoic acid, 3r-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylpentanoic acid, fmoc-beta-leu-oh, fmoc-l-, a-leucine, fmoc-l-beta-leucine, fmoc-, a-hoval-oh
PubChem CID 2761560
IUPAC Name (3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoic acid
SMILES CC(C)[C@@H](CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Quantity 250 mg
Beilstein 7388393
Solubility Information Slightly soluble in water.
Formula Weight 353.42
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-beta-homovaline
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only