missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-glycine, 98%

Catalog No. AAB2105003
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2105003 1 g
AAB2105006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AAB2105003 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' B2105003

CAS: 29022-11-5 | C17H15NO4 | 297.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 29022-11-5
Molecular Formula C17H15NO4
Molecular Weight (g/mol) 297.31
MDL Number MFCD00037140
InChI Key NDKDFTQNXLHCGO-UHFFFAOYSA-N
Synonym fmoc-gly-oh, fmoc-glycine, n-9h-fluoren-9-ylmethoxy carbonyl glycine, n-fmoc-glycine, fmoc-dl-glycine, n-9-fmoc-l-glycine, n-alpha-fmoc-glycine, fmoc-gly, n-9-fluorenylmethoxycarbonyl glycine
PubChem CID 93124
IUPAC Name 2-(9H-fluoren-9-ylmethoxycarbonylamino)acetic acid
SMILES OC(=O)CNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 172°C to 175°C
Quantity 1 g
Beilstein 2163967
Formula Weight 297.32
Percent Purity 98%
Chemical Name or Material N-Fmoc-glycine
EINECSNumber 249-373-3
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only