missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-D-allo-isoleucine, 98%
Description
N-Fmoc-D-allo-isoleucine used as a intermediate in pharmaceutical and chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Fmoc-D-allo-isoleucine used as a intermediate in pharmaceutical and chemical synthesis.
Solubility
Slightly soluble in water.
Notes
Refrigerate at -20°C. Keep container tightly closed. Store away from oxidizing agent.
N-Fmoc-D-allo-isoleucine used as a intermediate in pharmaceutical and chemical synthesis.
Solubility
Slightly soluble in water.
Notes
Refrigerate at -20°C. Keep container tightly closed. Store away from oxidizing agent.
Spécifications
Spécifications
| Solubility Information | Slightly soluble in water. |
| Quantity | 5 g |
| Formula Weight | 353.41 |
| Percent Purity | 98% |
| Physical Form | Solid |
| Chemical Name or Material | N-Fmoc-D-allo-isoleucine |
Sécurité et manipulation
| TSCA | No |
| missing translation for 'storageNote' | 2° to 8°C |
| Recommended Storage | Keep cold |
RUO – Research Use Only