missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-beta-alanine, 99%

Catalog No. AAH5988714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5988714 25 g
AAH5988706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5988714 Supplier Thermo Scientific Chemicals Supplier No. H5988714

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 35737-10-1 | C18H16NO4 | 310.33 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water and 1% acetic acid.

Notes
Desiccate at -20°C. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 35737-10-1
Molecular Formula C18H16NO4
Molecular Weight (g/mol) 310.33
MDL Number MFCD00063328
InChI Key LINBWYYLPWJQHE-UHFFFAOYSA-M
Synonym fmoc-beta-alanine, fmoc-beta-ala-oh, fmoc-b-ala-oh, fmoc-?-ala-oh, n-fmoc-beta-alanine, fmoc-b-alanine, 3-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-, a-ala-oh, 3-9h-fluoren-9-yl methoxy carbonyl amino propanoic acid, n-9h-fluoren-9-ylmethoxy carbonyl-beta-alanine
PubChem CID 2724630
IUPAC Name 3-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES [O-]C(=O)CCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 142°C to 147°C
Linear Formula FmocNHCH2CH2CO2H
Quantity 25 g
Beilstein 2302327
Solubility Information Soluble in water and 1% acetic acid.
Formula Weight 311.34
Percent Purity 99%
Chemical Name or Material N-Fmoc-beta-alanine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only