missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-4-(trifluoromethyl)-L-phenylalanine, 95%

Catalog No. AAH66549MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66549MD 250 mg
AAH6654903 1 g
AAH6654906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH66549MD Supplier Thermo Scientific Chemicals Supplier No. H66549MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 247113-86-6 | C25H20F3NO4 | 455.433 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 247113-86-6
Molecular Formula C25H20F3NO4
Molecular Weight (g/mol) 455.433
MDL Number MFCD00797581
InChI Key YMEGJWTUWMVZPD-QFIPXVFZSA-N
Synonym fmoc-phe 4-cf3-oh, fmoc-l-4-trifluoromethylphenylalanine, fmoc-4-trifluoromethyl-l-phenylalanine, fmoc-l-4-trifluoromethylphe, 4-trifluoromethyl-l-phenyl alanine, fmoc protected, 4-trifluoromethyl-l-phenylalanine, n-fmoc protected, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-4-trifluoromethyl phenyl propanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-trifluoromethyl phenyl propanoic acid, n-9h-fluoren-9-yl methoxy carbonyl-4-trifluoromethyl-l-phenylalanine
PubChem CID 7021069
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-(trifluoromethyl)phenyl]propanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)C(F)(F)F)C(=O)O

Specifications

Melting Point 127°C to 130°C
Color Brown
Quantity 250 mg
Formula Weight 455.43
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Fmoc-4-(trifluoromethyl)-L-phenylalanine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only