missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-4-iodo-L-phenylalanine, 95%

Catalog No. AAH5197706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5197706 5 g
AAH5197703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5197706 Supplier Thermo Scientific Chemicals Supplier No. H5197706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 82565-68-2 | C24H20INO4 | 513.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 82565-68-2
Molecular Formula C24H20INO4
Molecular Weight (g/mol) 513.33
MDL Number MFCD00672340
InChI Key LXOXXTQKKRJNNB-QFIPXVFZSA-N
Synonym fmoc-4-iodo-l-phenylalanine, fmoc-phe 4-i-oh, fmoc-l-4-iodophenylalanine, fmoc-l-phe 4-i-oh, fmoc-p-iodo-phe-oh, fmoc-4'-iodo-l-phe, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-iodophenyl propanoic acid, fmoc-l-phe 4-i, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-4-iodophenyl propanoic acid, l-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-4-iodo
PubChem CID 2761479
SMILES OC(=O)[C@H](CC1=CC=C(I)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 213°C to 217°C
Quantity 5 g
Sensitivity Light sensitive
Optical Rotation −10° (c=1 in DMF)
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-iodophenyl)propanoic acid
Formula Weight 513.33
Percent Purity 95%
Chemical Name or Material N-Fmoc-4-iodo-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only